Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Chordin-like 2/CHRDL2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB24481
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB24481 100 μg
MAB24481SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB24481 Supplier R&D Systems Supplier No. MAB24481
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Chordin-like 2/CHRDL2 Monoclonal specifically detects Chordin-like 2/CHRDL2 in Human samples. It is validated for Western Blot, Intracellular Staining by Flow Cytometry.

Specifications

Antigen Chordin-like 2/CHRDL2
Applications Western Blot, Flow Cytometry
Classification Monoclonal
Clone 272905
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. NP_056239
Gene Alias BNF-1, Chordin like 2, chordin-like 2, chordin-like protein 2, CHRDL2
Gene Symbols CHRDL2
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Chordin-like 2/CHRDL2 Arg28-Thr451 Accession # NP_056239
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25884
Test Specificity Detects human Chordin-like 2/CHRDL2 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant mouse CHRDL1 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.