Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CHRM2 Alexa Fluor™ 700-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB6377N100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB6377N100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB6377N100 Supplier R&D Systems Supplier No. FAB6377N100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Muscarinic Acetylcholine Receptor M2/CHRM2 Monoclonal antibody specifically detects Muscarinic Acetylcholine Receptor M2/CHRM2 in Human samples. It is validated for Flow Cytometry

Specifications

Antigen Muscarinic Acetylcholine Receptor M2/CHRM2
Applications Flow Cytometry
Classification Monoclonal
Clone 585713
Conjugate Alexa Fluor 700
Dilution Flow Cytometry 0.25-1 ug/106 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Gene Accession No. P08172
Gene Alias 7TM receptor, cholinergic receptor, muscarinic 2, FLJ43243, HM2, mAChR M2, MGC120006, MGC120007, muscarinic acetylcholine receptor M2, muscarinic M2 receptor
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human CHRM2, Accession # P08172
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1129
Reconstitution Reconstitution default not known
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.