Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CHRM2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB6377
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB6377 100 μg
MAB6377SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB6377 Supplier R&D Systems Supplier No. MAB6377
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Muscarinic Acetylcholine Receptor M2/CHRM2 Monoclonal specifically detects Muscarinic Acetylcholine Receptor M2/CHRM2 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, CyTOF-ready.

Specifications

Antigen Muscarinic Acetylcholine Receptor M2/CHRM2
Applications Flow Cytometry, Immunohistochemistry, CyTOF
Classification Monoclonal
Clone 585713
Concentration LYOPH
Conjugate Unconjugated
Dilution Flow Cytometry 0.25 ug/10^6 cells, Immunohistochemistry 8-25 ug/mL, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P08172
Gene Alias 7TM receptor, cholinergic receptor, muscarinic 2, FLJ43243, HM2, mAChR M2, MGC120006, MGC120007, muscarinic acetylcholine receptor M2, muscarinic M2 receptor
Gene Symbols CHRM2
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human CHRM2 Accession # P08172
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1129
Test Specificity Detects human CHRM2 in immunohistochemistry.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.