Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Chromogranin B Antibody, R&D Systems™

Mouse Monoclonal Antibody
$201.00 - $503.00
Specifications
| Antigen | Chromogranin B |
|---|---|
| Clone | 914332R |
| Dilution | ELISA |
| Applications | ELISA |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB88681SP
![]() |
R&D Systems
MAB88681SP |
25 μg |
Each for $201.00
|
|
|||||
MAB88681100
![]() |
R&D Systems
MAB88681100 |
100 μg |
Each for $503.00
|
|
|||||
Description
Chromogranin B Monoclonal specifically detects Chromogranin B in Human samples. It is validated for ELISA.Specifications
| Chromogranin B | |
| ELISA | |
| Monoclonal | |
| Supernatant | |
| Human | |
| P05060 | |
| 1114 | |
| IgG1 | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 914332R | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| CgB, CHGB, chromogranin B (secretogranin 1), Chromogranin-B, SCG1, SCG1sgI, secretogranin B, Secretogranin I, secretogranin-1, SgI | |
| Chinese hamster ovary cell line CHO-derived recombinant human Chromogranin B Met1-Gly677 Accession # P05060 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Chromogranin B in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title