Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Chromogranin B Antibody, R&D Systems™

Mouse Monoclonal Antibody
$201.00 - $503.00
Specifications
Antigen | Chromogranin B |
---|---|
Clone | 914316R |
Dilution | ELISA |
Applications | ELISA |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB88682SP
![]() |
R&D Systems
MAB88682SP |
25 μg |
Each for $201.00
|
|
|||||
MAB88682100
![]() |
R&D Systems
MAB88682100 |
100 μg |
Each for $503.00
|
|
|||||
Description
Chromogranin B Monoclonal specifically detects Chromogranin B in Human samples. It is validated for ELISA.Specifications
Chromogranin B | |
ELISA | |
Monoclonal | |
Supernatant | |
Human | |
P05060 | |
1114 | |
IgG1 | |
Protein A or G purified from cell culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
914316R | |
ELISA | |
Unconjugated | |
Mouse | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
CgB, CHGB, chromogranin B (secretogranin 1), Chromogranin-B, SCG1, SCG1sgI, secretogranin B, Secretogranin I, secretogranin-1, SgI | |
Chinese hamster ovary cell line CHO-derived recombinant human Chromogranin B Met1-Gly677 Accession # P05060 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human Chromogranin B in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title