Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Chromogranin B Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB8868
This item is not returnable.
View return policy
Description
Chromogranin B Monoclonal specifically detects Chromogranin B in Human samples. It is validated for Immunohistochemistry.Specifications
Chromogranin B | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CgB, CHGB, chromogranin B (secretogranin 1), Chromogranin-B, SCG1, SCG1sgI, secretogranin B, Secretogranin I, secretogranin-1, SgI | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
1114 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Immunohistochemistry | |
914334 | |
Immunohistochemistry 8-25 ug/mL | |
P05060 | |
CHGB | |
Chinese hamster ovary cell line CHO-derived recombinant human Chromogranin B Met1-Gly677 Accession # P05060 | |
100 μg | |
Primary | |
Detects human Chromogranin B in direct ELISAs. | |
Human | |
Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction