Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Chymase/CMA1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB4099SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB4099SP 25 μg
MAB4099 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB4099SP Supplier R&D Systems Supplier No. MAB4099SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Chymase/CMA1/Mast Cell Chymase Monoclonal specifically detects Chymase/CMA1/Mast Cell Chymase in Human samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen Chymase/CMA1/Mast Cell Chymase
Applications Western Blot, Immunoprecipitation
Classification Monoclonal
Clone 422818
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P23946
Gene Alias Alpha-chymase, chymase, chymase 1 preproprotein transcript E, chymase 1 preproprotein transcript I, chymase 1, mast cell, chymase, heart, chymase, mast cell, CMA1, CYH, CYM, EC 3.4.21, EC 3.4.21.39, Mast cell protease I, MCP3P, Mcpt5, MCT1, MGC119890, MGC119891
Gene Symbols CMA1
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Chymase/CMA1 Gly20-Asn247 Accession # P23946
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1215
Test Specificity Detects human Chymase/CMA1 in direct ELISAs and Western blots. Does not cross-react with recombinant mouse (rm) MCPT-1, rmMCPT-6, rmMCPT-7, or rmMCPT-11.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.