Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human cIAP-1/HIAP-2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB818
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB818 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB818 Supplier R&D Systems Supplier No. MAB818
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

cIAP-1/HIAP-2 Monoclonal specifically detects cIAP-1/HIAP-2 in Human samples. It is validated for Western Blot, Simple Western, Knockout Validated.

Specifications

Antigen cIAP-1/HIAP-2
Applications Western Blot, KnockDown
Classification Monoclonal
Clone 681732
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Knockout Validated
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q13490
Gene Alias API1Hiap-2, apoptosis inhibitor 1, baculoviral IAP repeat containing 2, baculoviral IAP repeat-containing 2, baculoviral IAP repeat-containing protein 2, BIRC2, cIAP1, C-IAP1, hIAP2, HIAP-2, IAP homolog B, IAP2, IAP-2, Inhibitor of apoptosis protein 2, MIHB, MIHBHIAP2, NFR2-TRAF signalling complex protein, RING finger protein 48, RNF48hiap-2, TNFR2-TRAF-signaling complex protein 2
Gene Symbols BIRC2
Host Species Mouse
Immunogen E. coli-derived recombinant human cIAP-1/HIAP-2 His2-Ser618 Accession # Q13490
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 329
Test Specificity Detects human cIAP-1/HIAP-2 in direct ELISAs and Western blots. In direct ELISAs, 100% cross-reactivity with recombinant human (rh) cIAP-2 (aa 2-604) and no cross-reactivity with rhBIRC6 (aa 4582-4735), rhcIAP-2 (aa 94-178), rhXIAP (aa 1-497), rhXIAP (BIR2 domain; aa 124-242), or rhXIAP (BIR3 domain; aa 252-356) is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.