Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human cIAP-2/HIAP-1 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 11 publications
Supplier: R&D Systems AF8171
This item is not returnable.
View return policy
Description
cIAP-2/HIAP-1 Polyclonal specifically detects cIAP-2/HIAP-1 in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry.Specifications
cIAP-2/HIAP-1 | |
Polyclonal | |
Unconjugated | |
U45878 | |
BIRC3 | |
E. coli-derived recombinant human cIAP-2/HIAP-1 Asn2-Ser604 Accession # U45878 | |
100 μg | |
Primary | |
Detects human cIAP-2/HIAP-1 in Western blots. In Western blots, less than 1% cross-reactivity with cIAP-1 is observed. | |
Human | |
IgG |
Western Blot, Immunohistochemistry | |
LYOPH | |
Western Blot 0.5 μg/mL, Simple Western 5 μg/mL, Immunohistochemistry 1-15 μg/mL | |
AIP1, API2, BIRC3, BIRC3 baculoviral IAP repeat containing 3, c-IAP2, CIAP2, HAIP1, HIAP1, HIAP-1, MALT2, MIHC, RNF49 | |
Goat | |
Affinity Purified | |
RUO | |
330 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction