Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CILP-1 C-Terminal Fragment Antibody, R&D Systems™

Sheep Polyclonal Antibody
$200.50 - $502.50
Specifications
Antigen | CILP-1 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
CILP-1 Polyclonal specifically detects CILP-1 in Human samples. It is validated for Western Blot.Specifications
CILP-1 | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Sheep | |
Human | |
cartilage intermediate layer protein 1, cartilage intermediate layer protein 1 C1, cartilage intermediate layer protein 1 C2, cartilage intermediate layer protein, nucleotide pyrophosphohydrolase, Cartilage intermediate-layer protein, CILP, CILP1, CILP-1, HsT18872 | |
CILP | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
O75339 | |
8483 | |
Mouse myeloma cell line NS0-derived recombinant human CILP-1 C-Terminal Fragment Arg724-Asn1184 Accession # O75339 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human CILP-1 C-Terminal Fragment in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human CILP-1 N-terminal peptide is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title