Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CIN85/SH3KBP1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB8877
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB8877 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB8877 Supplier R&D Systems Supplier No. MAB8877
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

CIN85/SH3KBP1 Monoclonal specifically detects CIN85/SH3KBP1 in Human samples. It is validated for Western Blot.

Specifications

Antigen CIN85/SH3KBP1
Applications Western Blot
Classification Monoclonal
Clone 931308
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q96B97
Gene Alias c-Cbl-interacting protein, CD2-binding protein 3, CD2BP3, CIN85Cbl-interacting protein of 85 kDa, GIG10, HSB1, HSB-1, Human Src family kinase-binding protein 1, MIG18, migration-inducing gene 18, SH3 domain-containing kinase-binding protein 1, SH3-domain kinase binding protein 1, SH3KBP1, Src family kinase-binding protein 1, src-related kinase binding protein-1
Gene Symbols SH3KBP1
Host Species Mouse
Immunogen E. coli-derived recombinant human CIN85/SH3KBP1 Pro366-Lys665 Accession # Q96B97
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 30011
Test Specificity Detects human CIN85/SH3KBP1 in direct ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.