Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CL-P1/COLEC12 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB26901SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB26901SP 25 μg
MAB26901 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB26901SP Supplier R&D Systems Supplier No. MAB26901SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

CL-P1/COLEC12 Monoclonal specifically detects CL-P1/COLEC12 in Human samples. It is validated for Western Blot.

Specifications

Antigen CL-P1/COLEC12
Applications Western Blot
Classification Monoclonal
Clone 388305
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q5KU26
Gene Alias CL-P1, CLP1hCL-P1, COLEC12, collectin placenta 1, Collectin placenta protein 1, collectin sub-family member 12, collectin-12, NSR2, Nurse cell scavenger receptor 2, SCARA4, SCARA4NSR2, Scavenger receptor class A member 4, scavenger receptor class A, member 4, SRCL Type I, SRCLScavenger receptor with C-type lectin
Gene Symbols COLEC12
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant human CL-P1/COLEC12 isoform 1 Leu57-Leu742 Accession # Q5KU26
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81035
Test Specificity Detects human CL-P1/COLEC12 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse CL-P1 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.