Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CL-P1/COLEC12 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF2690
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF2690 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF2690 Supplier R&D Systems Supplier No. AF2690
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 4 publications

CL-P1/COLEC12 Polyclonal specifically detects CL-P1/COLEC12 in Human samples. It is validated for Western Blot, Flow Cytometry, Blockade of Receptor-ligand Interaction, Immunocytochemistry, CyTOF-ready.

Specifications

Antigen CL-P1/COLEC12
Applications Western Blot, Flow Cytometry, Blockade of Receptor-ligand Interaction, Immunocytochemistry, CyTOF
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Blockade of Receptor-ligand Interaction, Immunocytochemistry 5-15 ug/mL, CyTOF-ready
Gene Accession No. Q5KU26
Gene Alias CL-P1, CLP1hCL-P1, COLEC12, collectin placenta 1, Collectin placenta protein 1, collectin sub-family member 12, collectin-12, NSR2, Nurse cell scavenger receptor 2, SCARA4, SCARA4NSR2, Scavenger receptor class A member 4, scavenger receptor class A, member 4, SRCL Type I, SRCLScavenger receptor with C-type lectin
Gene Symbols COLEC12
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human CL-P1/COLEC12 isoform 1 Leu57-Leu742 Accession # Q5KU26
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81035
Test Specificity Detects CL-P1/COLEC12 in direct ELISAs and Western blots. In direct ELISAs, approximately 45% cross-reactivity with recombinant mouse CL-P1 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.