Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Claspin Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAF3310
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAF3310 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAF3310 Supplier R&D Systems Supplier No. BAF3310
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Claspin Polyclonal specifically detects Claspin in Human samples. It is validated for Western Blot.

Specifications

Antigen Claspin
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot 0.1 ug/mL
Gene Accession No. Q9HAW4
Gene Alias claspin, claspin homolog, claspin homolog (Xenopus laevis), CLSPN, hClaspin, MGC131612, MGC131613, MGC131615
Gene Symbols CLSPN
Host Species Goat
Immunogen E. coli-derived recombinant human Claspin Phe1138-Ser1339 Accession # Q9HAW4
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 63967
Test Specificity Detects human Claspin in Western blots.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.