Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Claudin-1 Alexa Fluor 594-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB4618T100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB4618T100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB4618T100 Supplier R&D Systems Supplier No. FAB4618T100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

Claudin-1 Monoclonal specifically detects Claudin-1 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen Claudin-1
Applications Flow Cytometry
Classification Monoclonal
Clone 421203
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 594
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. O95832
Gene Alias claudin 1, claudin-1, Claudin1, CLD1, CLDN1, ILVASCSenescence-associated epithelial membrane protein, SEMP1, SEMP1senescence-associated epithelial membrane protein 1
Gene Symbols CLDN1
Host Species Rat
Immunogen CHO Chinese hamster ovary cell line transfected with human Claudin-1 Met1-Val211 Accession # O95832
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9076
Test Specificity Stains human Claudin-1 transfectants but not irrelevant transfectants.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2a
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.