Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Claudin-1 APC-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems FAB4618A
This item is not returnable.
View return policy
Description
Claudin-1 Monoclonal specifically detects Claudin-1 in Human samples. It is validated for Flow Cytometry.Specifications
Claudin-1 | |
Monoclonal | |
APC | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
claudin 1, claudin-1, Claudin1, CLD1, CLDN1, ILVASCSenescence-associated epithelial membrane protein, SEMP1, SEMP1senescence-associated epithelial membrane protein 1 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
9076 | |
Human | |
Purified |
Flow Cytometry | |
421203 | |
Flow Cytometry 10 uL/10^6 cells | |
O95832 | |
CLDN1 | |
CHO Chinese hamster ovary cell line transfected with human Claudin-1 Met1-Val211 Accession # O95832 | |
100 Tests | |
Primary | |
Stains human Claudin-1 transfectants but not irrelevant transfectants. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction