Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Claudin-3 APC-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB4620A
This item is not returnable.
View return policy
Description
Claudin-3 Monoclonal specifically detects Claudin-3 in Human samples. It is validated for Flow Cytometry.Specifications
| Claudin-3 | |
| Monoclonal | |
| APC | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| C7orf1, claudin 3, claudin-3, Claudin3, CLDN3, Clostridium perfringens enterotoxin receptor 2, CPE-R 2, CPE-R2, CPETR2, CPETR2CPE-receptor 2, HRVP1, Rat ventral prostate.1 protein homolog, RVP1, ventral prostate.1 protein homolog, ventral prostate.1-like protein | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 1365 | |
| Human | |
| Purified |
| Flow Cytometry | |
| 385021 | |
| Flow Cytometry 10 uL/10^6 cells | |
| O15551 | |
| CLDN3 | |
| HEK293 human embryonic kidney cell line transfected with human Claudin-3 Met1-Val220 Accession # O15551 | |
| 100 Tests | |
| Primary | |
| Stains human Claudin-3 transfectants but not irrelevant transfectants. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction