Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Claudin-4 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB42191SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB42191SP 25 μg
MAB42191100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB42191SP Supplier R&D Systems Supplier No. MAB42191SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Claudin-4 Monoclonal specifically detects Claudin-4 in Human samples. It is validated for Immunocytochemistry.

Specifications

Antigen Claudin-4
Applications Immunocytochemistry
Classification Monoclonal
Clone 382316
Conjugate Unconjugated
Dilution Immunocytochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. NP_001296
Gene Alias claudin 4, Claudin4, CLDN4, Clostridium perfringens enterotoxin receptor, Clostridium perfringens enterotoxin receptor 1, CPE-R, CPE-RCPETR, CPE-receptor, CPETR1, hCPE-R, WBSCR8, WBSCR8CPERCPETR1claudin-4, Williams-Beuren syndrome chromosomal region 8 protein
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human Claudin-4, Met1-Val209, Accession No. NP_001296
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Primary or Secondary Primary
Gene ID (Entrez) 1364.0
Test Specificity Detects human Claudin-4 in direct ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.