Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Claudin-6 Alexa Fluor 750-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB3656S100UG
This item is not returnable.
View return policy
Description
Claudin-6 Monoclonal specifically detects Claudin-6 in Human samples. It is validated for Flow Cytometry.Specifications
Claudin-6 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
P56747 | |
CLDN6 | |
NS0 mouse myeloma cell line transfected with human Claudin-6 Met1-Val220 Accession # P56747 | |
100 μg | |
Primary | |
Stains human Claudin-6 transfected cells but not irrelevant transfectants. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Flow Cytometry | |
342927 | |
Alexa Fluor 750 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
claudin 6, claudin-6, Claudin6, CLDN6, skullin, Skullin 2 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
9074 | |
Human | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction