Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Claudin-6 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB3656SP
This item is not returnable.
View return policy
Description
Claudin-6 Monoclonal specifically detects Claudin-6 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOF-ready.Specifications
Claudin-6 | |
Monoclonal | |
LYOPH | |
Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 8-25 ug/mL, CyTOF-ready | |
P56747 | |
CLDN6 | |
NS0 mouse myeloma cell line transfected with human Claudin-6 Met1-Val220 Accession # P56747 | |
25 μg | |
Primary | |
Stains human Claudin-6 transfected cells but not irrelevant transfectants. | |
Human | |
Purified |
Flow Cytometry, Immunocytochemistry, CyTOF | |
342927 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
claudin 6, claudin-6, Claudin6, CLDN6, skullin, Skullin 2 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
9074 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction