Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Claudin-8 Alexa Fluor 647-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. IC5275R100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC5275R100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC5275R100 Supplier R&D Systems Supplier No. IC5275R100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Claudin-8 Monoclonal specifically detects Claudin-8 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.

Specifications

Antigen Claudin-8
Applications Flow Cytometry
Classification Monoclonal
Clone 474210
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 647
Dilution Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. P56748
Gene Alias claudin 8, Claudin8, CLDN8, human CLDN8 gene for claudin-810claudin-8
Gene Symbols CLDN8
Host Species Mouse
Immunogen HEK293 human embryonic kidney cell line transfected with human Claudin-8 Met1-Val225 (Ser151Pro) Accession # P56748
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9073
Test Specificity Detects human Claudin-8. Stains human Claudin-8 transfectants but not irrelevant transfectants.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.