Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CLC Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAF962
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAF962 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAF962 Supplier R&D Systems Supplier No. BAF962
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

CLC Polyclonal specifically detects CLC in Human samples. It is validated for Western Blot.

Specifications

Antigen CLC
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot 0.1 ug/mL
Gene Accession No. Q9UBD9
Gene Alias B-cell stimulating factor 3, B-cell stimulatory factor 3, BSF-3, BSF3BSF-3, cardiotrophin-like cytokine factor 1, CISS2, CLCF1, CLCNNT-1, CRLF1 associated cytokine-like factor 1, neurotrophin-1/B-cell stimulating factor-3, NNT-1, NNT1B-cell-stimulating factor 3, Novel neurotrophin-1, NR6
Gene Symbols CLCF1
Host Species Goat
Immunogen E. coli-derived recombinant human CLC Leu28-Phe225 Accession # Q9UBD9
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23529
Test Specificity Detects human CLC in Western blots. In Western blots, less than 1% cross-reactivity with recombinant human CT-1 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.