Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CLEC-2A Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB72191
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB72191 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB72191 Supplier R&D Systems Supplier No. MAB72191
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

CLEC-2A Monoclonal specifically detects CLEC-2A in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen CLEC-2A
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone 705529
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.5 ug/mL, Immunohistochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q6UVW9
Gene Alias CLEC2A, CLEC2A C-type lectin domain family 2, member A, KACL, PILAR, UNQ5792
Gene Symbols CLEC2A
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human CLEC-2A Trp49-Leu174 (predicted) Accession # Q6UVW9
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 387836
Test Specificity Detects human CLEC-2A in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human (rh) CLEC-2D, rhCLEC-2, or rhCD69 is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.