Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CLEC10A/CD301 Alexa Fluor 488-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB48881G
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB48881G 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB48881G Supplier R&D Systems Supplier No. FAB48881G
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

CLEC10A/CD301 Monoclonal specifically detects CLEC10A/CD301 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen CLEC10A/CD301
Applications Flow Cytometry
Classification Monoclonal
Clone 744812
Conjugate Alexa Fluor 488
Dilution Flow Cytometry 5 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. Q8IUN9
Gene Alias CD301, CD301 antigen, CD301C-type lectin domain family 10 member A, CLECSF13, CLECSF14, CLECSF14macrophage C-type lectin, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 14 (macrophage-derived), C-type lectin domain family 10, member A, C-type lectin superfamily member 14, HML2, HMLC-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 13 (macrophage-derived), Macrophage lectin 2, macrophage lectin 2 (calcium dependent), MGL
Gene Symbols CLEC10A
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human CLEC10A/CD301 Gln61-His316 Accession # Q8IUN9
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10462
Test Specificity Detects human CLEC10A/CD301 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human ASGPR1 or recombinant mouse MGL1 is observed.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.