Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CLEC14A Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAF4968
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAF4968 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAF4968 Supplier R&D Systems Supplier No. BAF4968
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

CLEC14A Polyclonal specifically detects CLEC14A in Human samples. It is validated for Western Blot.

Specifications

Antigen CLEC14A
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot 0.1 ug/mL
Gene Accession No. Q86T13
Gene Alias C14orf27, CEG1, chromosome 14 open reading frame 27, ClECT and EGF-like domain containing protein, C-type lectin domain family 14 member A, C-type lectin domain family 14, member A, EGF R5, EGFR5, EGFR-5, Epidermal growth factor receptor 5
Gene Symbols CLEC14A
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant human CLEC14A Glu22-Ala397 Accession # Q86T13
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 161198
Test Specificity Detects human CLEC14A in Western blots. In Western blots, less than 1% cross-reactivity with recombinant human (rh) CLEC1, rhCLEC2, rhCLEC3B, and rhCLEC10A is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.