Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CLEC17A Alexa Fluor™ 488-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB101471G100UG
This item is not returnable.
View return policy
Description
CLEC17A Monoclonal antibody specifically detects CLEC17A in Human samples. It is validated for Flow CytometrySpecifications
| CLEC17A | |
| Monoclonal | |
| Alexa Fluor 488 | |
| Q6ZS10 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 388512 | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2°C to 8°C as supplied. | |
| IgG1 |
| Flow Cytometry | |
| 1064719 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| C-type lectin domain family 17, member A | |
| CHO-derived human CLEC17A, Lys194-Cys378, Accession # Q6ZS10 | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction