Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CLEC17A Alexa Fluor™ 594-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB101471T100UG
This item is not returnable.
View return policy
Description
CLEC17A Monoclonal antibody specifically detects CLEC17A in Human samples. It is validated for Flow CytometrySpecifications
CLEC17A | |
Monoclonal | |
Alexa Fluor 594 | |
Q6ZS10 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
388512 | |
Protect from light. Do not freeze. 12 months from date of receipt, 2°C to 8°C as supplied. | |
IgG1 |
Flow Cytometry | |
1064719 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
C-type lectin domain family 17, member A | |
CHO-derived human CLEC17A, Lys194-Cys378, Accession # Q6ZS10 | |
100 μg | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction