Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CLEC17A Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. RD10000121
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RD10000121 100 μg
MAB101471SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. RD10000121 Supplier R&D Systems Supplier No. MAB101471100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

CLEC17A Monoclonal antibody specifically detects CLEC17A in Human samples. It is validated for Flow Cytometry

Specifications

Antigen CLEC17A
Applications Flow Cytometry
Classification Monoclonal
Clone 1064719
Conjugate Unconjugated
Dilution 0.25 μg/106 cells
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS.
Gene Accession No. Q6ZS10
Gene Alias C-type lectin domain family 17, member A
Host Species Mouse
Immunogen CHO-derived human CLEC17A, Lys194-Cys378, Accession # Q6ZS10
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 388512
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 6 months, -20°C to -70°C under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.