Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CLEC3B/Tetranectin Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF5170SP
This item is not returnable.
View return policy
Description
CLEC3B/Tetranectin Polyclonal specifically detects CLEC3B/Tetranectin in Human samples. It is validated for Western Blot.Specifications
CLEC3B/Tetranectin | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
C-type lectin domain family 3 member B, C-type lectin domain family 3, member B, DKFZp686H17246, Plasminogen kringle 4-binding protein, TETN, Tetranectin, tetranectin (plasminogen binding protein), TNA, TNAtetranectin (plasminogen-binding protein), TNtetranectin | |
Sheep | |
Affinity Purified | |
RUO | |
7123 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
P05452 | |
CLEC3B | |
Mouse myeloma cell line NS0-derived recombinant human CLEC3B/Tetranectin Glu22-Val202 Accession # P05452 | |
25 μg | |
Primary | |
Detects human CLEC3B/Tetranectin in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 5% cross-reactivity with recombinant human (rh) CLEC1, rhCLEC2, rhCLEC10A, and rhCLEC14A is observed. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction