Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CLEC4D/CLECSF8 Alexa Fluor 750-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB2806S100UG
This item is not returnable.
View return policy
Description
CLEC4D/CLECSF8 Monoclonal specifically detects CLEC4D/CLECSF8 in Human samples. It is validated for Flow Cytometry.Specifications
CLEC4D/CLECSF8 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
Q8WXI8 | |
CLEC4D | |
Mouse myeloma cell line NS0-derived recombinant human CLEC4D/CLECSF8 Gly52-Asn215 Accession # Q8WXI8 | |
RUO | |
338339 | |
Human | |
IgG2b |
Flow Cytometry | |
413512 | |
Alexa Fluor 750 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
CD368, CLEC6, CLEC-6, CLECSF8, CLECSF8Mpcl, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 8, C-type lectin domain family 4 member D, C-type lectin domain family 4, member D, C-type lectin receptor, C-type lectin superfamily member 8, C-type lectin-like receptor 6, macrophage C-type lectin, MCL, MGC40078, MPCL | |
Mouse | |
100 μg | |
Primary | |
Detects human CLEC4D/CLECSF8 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) CLECSF9, rhCLECSF13, rhOCIL, or rhOCILrp2 is observed. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction