Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CLEC4E Alexa Fluor 700-conjugated Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Supplier: R&D Systems FAB8995N100UG
This item is not returnable.
View return policy
Description
CLEC4E Monoclonal specifically detects CLEC4E in Human samples. It is validated for Flow Cytometry.Specifications
CLEC4E | |
Monoclonal | |
Alexa Fluor 700 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
CLECSF9, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 9, C-type lectin domain family 4 member E, C-type lectin domain family 4, member E, Macrophage-inducible C-type lectin, MINCLE, MINCLEC-type lectin superfamily member 9 | |
Chinese Hamster Ovary cell line, CHO-derived recombinant human CLEC4E, Arg41-Leu219, Accession # Q9ULY5 | |
100 μg | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
2455C | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
Q9ULY5 | |
Rabbit | |
Protein A or G purified from cell culture supernatant | |
RUO | |
26253 | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction