Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CLEC4E Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB8995SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB8995SP 25 μg
MAB8995100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB8995SP Supplier R&D Systems Supplier No. MAB8995SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

CLEC4E Monoclonal specifically detects CLEC4E in Human samples. It is validated for Flow Cytometry, CyTOF-ready.

Specifications

Antigen CLEC4E
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 2455C
Conjugate Unconjugated
Dilution Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready
Formulation Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS.
Gene Accession No. Q9ULY5
Gene Alias CLECSF9, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 9, C-type lectin domain family 4 member E, C-type lectin domain family 4, member E, Macrophage-inducible C-type lectin, MINCLE, MINCLEC-type lectin superfamily member 9
Host Species Rabbit
Immunogen Chinese Hamster Ovary cell line, CHO-derived recombinant human CLEC4E, Arg41-Leu219, Accession # Q9ULY5
Purification Method Protein A or G purified from cell culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26253
Test Specificity Detects human CLEC4E in direct ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.