Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CLEC9a Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB6049
This item is not returnable.
View return policy
Description
CLEC9a Monoclonal specifically detects CLEC9a in Human samples. It is validated for Flow Cytometry, Binding Inhibition, CyTOF-ready.Specifications
| CLEC9a | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD370, C-type lectin domain family 9 member A, C-type lectin domain family 9, member A, DNGR1, HEEE9341, UNQ9341 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 283420 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Flow Cytometry, CyTOF | |
| 683409 | |
| Flow Cytometry 2.5 ug/10^6 cells, Binding Inhibition 1 ug/mL, CyTOF-ready | |
| Q6UXN8 | |
| CLEC9A | |
| Mouse myeloma cell line NS0-derived recombinant human CLEC9a Lys57-Val241 Accession # Q6UXN8 | |
| 500 μg | |
| Primary | |
| Detects human CLEC9a in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) CLEC1, 2, 2A, 3B, 10A, 12B, 14A, rhCD302/CLEC13a, rhMICL, or recombinant mouse CLEC9a is observed. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction