Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CLP24 Alexa Fluor 594-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB6747T100UG
This item is not returnable.
View return policy
Description
CLP24/TMEM204 Monoclonal specifically detects CLP24/TMEM204 in Human samples. It is validated for Flow Cytometry.Specifications
CLP24/TMEM204 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
Q9BSN7 | |
TMEM204 | |
NS0 mouse myeloma cell line transfected with human CLP24 Accession # Q9BSN7 | |
RUO | |
79652 | |
Human | |
IgG2b |
Flow Cytometry | |
652424 | |
Alexa Fluor 594 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
C16orf30, C16orf30chromosome 16 open reading frame 30, Claudin-like protein 24, CLP24MGC111564, FLJ20898, TMEM204, transmembrane protein 204 | |
Mouse | |
100 μg | |
Primary | |
Detects human CLP24 in ELISAs. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction