Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Clusterin Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $467.50
Specifications
Antigen | Clusterin/APOJ |
---|---|
Clone | 350207 |
Dilution | ELISA |
Applications | ELISA |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB29373SP
![]() |
R&D Systems
MAB29373SP |
25 μg |
Each for $200.50
|
|
|||||
MAB29373100
![]() |
R&D Systems
MAB29373100 |
100 μg |
Each for $467.50
|
|
|||||
Description
Clusterin/APOJ Monoclonal specifically detects Clusterin/APOJ in Human samples. It is validated for ELISA.Specifications
Clusterin/APOJ | |
ELISA | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS. | |
40, 40, sulfated glycoprotein 2, Aging-associated gene 4 protein, aging-associated protein 4, apo-J, APOJ, Apolipoprotein J, CLI, CLIclusterin (complement lysis inhibitor, SP-40, CLU, clusterin, Complement cytolysis inhibitor, complement lysis inhibitor, Complement-associated protein SP-40, Ku70-binding protein 1, KUB1SGP2, MGC24903, NA1/NA2, SGP-2, SP-40, sulfated glycoprotein 2, Testosterone-repressed prostate message 2, testosterone-repressed prostate message 2, apolipoprotein J), TRPM-2, TRPM-2TRPM2 | |
Mouse myeloma cell line NS0-derived recombinant human Clusterin, Asp23-Arg227 (beta) & Ser228-Glu449 (alpha), Accession # NP_001822 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
350207 | |
ELISA | |
Unconjugated | |
Mouse | |
Human | |
NP_001822 | |
1191 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title