Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Clusterin-like 1/CLUL1 Antibody, R&D Systems™

Goat Polyclonal Antibody
$176.44 - $532.55
Specifications
| Antigen | Clusterin-like 1/CLUL1 |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.1 ug/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
Clusterin-like 1/CLUL1 Polyclonal specifically detects Clusterin-like 1/CLUL1 in Human samples. It is validated for Western Blot.Specifications
| Clusterin-like 1/CLUL1 | |
| Western Blot 0.1 ug/mL | |
| Polyclonal | |
| Goat | |
| Human | |
| CLUL1, Clusterin like 1, clusterin-like 1 (retinal), clusterin-like protein 1, RA337M, Retinal-specific clusterin-like protein | |
| CLUL1 | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Q15846 | |
| 27098 | |
| Mouse myeloma cell line NS0-derived recombinant human Clusterin-like 1/CLUL1 Cys20-Ala442 Accession # Q15846 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects human Clusterin-like 1/CLUL1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 5% cross-reactivity with recombinant human Clusterin is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title