Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CMG-2/ANTXR2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB29401SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB29401SP 25 μg
MAB29401100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB29401SP Supplier R&D Systems Supplier No. MAB29401SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

CMG-2/ANTXR2 Monoclonal specifically detects CMG-2/ANTXR2 in Human samples. It is validated for ELISA.

Specifications

Antigen CMG-2/ANTXR2
Applications ELISA
Classification Monoclonal
Clone 406510
Conjugate Unconjugated
Dilution ELISA
Formulation Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS.
Gene Accession No. P58335
Gene Alias anthrax toxin receptor 2, ANTXR2, Capillary morphogenesis gene 2 protein, capillary morphogenesis protein 2, cI-35, CMG2, CMG2MGC111533, CMG-2MGC45856, FLJ31074, ISH, JHF, JHS
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human CMG-2/ANTXR2 Ser22-Arg200 Accession # P58335
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Primary or Secondary Primary
Gene ID (Entrez) 118429
Test Specificity Detects human CMG-2/ANTXR2 in direct ELISAs.
Reconstitution Reconstitute at 0.5mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.