Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CMG-2/ANTXR2 APC-conjugated Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems FAB2940A
This item is not returnable.
View return policy
Description
CMG-2/ANTXR2 Polyclonal specifically detects CMG-2/ANTXR2 in Human samples. It is validated for Flow Cytometry.Specifications
CMG-2/ANTXR2 | |
Polyclonal | |
Flow Cytometry 10 uL/10^6 cells | |
anthrax toxin receptor 2, ANTXR2, Capillary morphogenesis gene 2 protein, capillary morphogenesis protein 2, cI-35, CMG2, CMG2MGC111533, CMG-2MGC45856, FLJ31074, ISH, JHF, JHS | |
Goat | |
Affinity Purified | |
RUO | |
118429 | |
Human | |
IgG |
Flow Cytometry | |
APC | |
P58335 | |
ANTXR2 | |
Mouse myeloma cell line NS0-derived recombinant human CMG-2/ANTXR2 isoform 1 Gln34-Asn317 Accession # P58335 | |
100 Tests | |
Primary | |
Detects human CMG-2/ANTXR2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 20% cross-reactivity with recombinant mouse CMG-2 is observed. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction