Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CNP Propeptide Antibody, R&D Systems™

Rat Monoclonal Antibody
$200.50 - $491.00
Specifications
| Antigen | CNP/NPPC |
|---|---|
| Clone | 347625 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL |
| Applications | Western Blot, Immunohistochemistry |
Description
CNP/NPPC Monoclonal specifically detects CNP/NPPC in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
| CNP/NPPC | |
| LYOPH | |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| Rat | |
| Human | |
| P23582 | |
| 4880 | |
| E. coli-derived recombinant human CNP Lys24-Cys126 Accession # P23582 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human CNP Propeptide in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-react with recombinant human (rh) ANP precursor or rhBNP precursor is observed. |
| 347625 | |
| Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CNP, CNP2, C-type natriuretic peptide, natriuretic peptide C, natriuretic peptide precursor C, NPPC | |
| NPPC | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title