Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CNTF Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF257NA
Change view
Click to view available options
Quantity:
1 mg
100 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF257NA 100 μg
AB257NA 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF257NA Supplier R&D Systems Supplier No. AF257NA
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 4 publications

CNTF Polyclonal specifically detects CNTF in Human samples. It is validated for Western Blot, Neutralization.

Specifications

Antigen CNTF
Applications Western Blot, Neutralization
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Neutralization 3-8 ug/mL
Gene Accession No. P26441
Gene Alias ciliary neurotrophic factor, HCNTF
Gene Symbols CNTF
Host Species Goat
Immunogen E. coli-derived recombinant human CNTF (R&D Systems, Catalog # 257-NT) Ala2-Met200 Accession # P26441
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1270
Test Specificity Detects human CNTF in direct ELISAs and Western blots. In direct ELISAs, approximately 100% cross-reactivity with recombinant rat CNTF is observed and less than 1% cross-reactivity with rhCNTF R alpha is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.