Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Coagulation Factor III/ Tissue Factor Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF2339SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF2339SP 25 μg
AF2339 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF2339SP Supplier R&D Systems Supplier No. AF2339SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 5 publications

Coagulation Factor III/Tissue Factor Polyclonal specifically detects Coagulation Factor III/Tissue Factor in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, CyTOF-ready.

Specifications

Antigen Coagulation Factor III/Tissue Factor
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, CyTOF
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.5 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Immunohistochemistry 5-15 ug/mL, Immunoprecipitation 25 ug/mL, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P13726
Gene Alias CD142, CD142 antigen, Coagulation factor III, coagulation factor III (thromboplastin, tissue factor), F3, FLJ17960, TF, TFA, Thromboplastin, tissue factor
Gene Symbols F3
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human Coagulation Factor III/Tissue Factor Gly34-Glu251 Accession # P13726
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2152
Test Specificity Detects human Coagulation Factor III/Tissue Factor in direct ELISAs and Western blots. In direct ELISAs, less than 5% cross-reactivity with recombinant mouse Coagulation Factor III/Tissue Factor is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.