Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Coagulation Factor VII Alexa Fluor 350-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB23381U100UG
This item is not returnable.
View return policy
Description
Coagulation Factor VII Monoclonal specifically detects Coagulation Factor VII in Human samples. It is validated for Flow Cytometry.Specifications
Coagulation Factor VII | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
NP_062562 | |
F7 | |
Mouse myeloma cell line NS0-derived recombinant human Coagulation Factor VII Ala39-Pro444 Accession # NP_062562 | |
RUO | |
2155 | |
Human | |
IgG2b |
Flow Cytometry | |
321605 | |
Alexa Fluor 350 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
coagulation factor VII, coagulation factor VII (serum prothrombin conversion accelerator), EC 3.4.21, EC 3.4.21.21, Eptacog alfa, F7, FVII coagulation protein, Proconvertin, Serum prothrombin conversion accelerator, SPCA | |
Mouse | |
100 μg | |
Primary | |
Detects human Coagulation Factor VII in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) Kallikrein-1, -2, -3, -4, -5, -7, -8, -9, -10, -11, -12, -13, -14, -15, -B1, rhHGFA, rhFactor X, rhFactor XI, rhThrombin, rhuPA, or rhPROC is observed. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction