Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Coagulation Factor VII Alexa Fluor 405-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FA23381V100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FA23381V100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FA23381V100 Supplier R&D Systems Supplier No. FAB23381V100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Coagulation Factor VII Monoclonal specifically detects Coagulation Factor VII in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen Coagulation Factor VII
Applications Flow Cytometry
Classification Monoclonal
Clone 321605
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 405
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. NP_062562
Gene Alias coagulation factor VII, coagulation factor VII (serum prothrombin conversion accelerator), EC 3.4.21, EC 3.4.21.21, Eptacog alfa, F7, FVII coagulation protein, Proconvertin, Serum prothrombin conversion accelerator, SPCA
Gene Symbols F7
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Coagulation Factor VII Ala39-Pro444 Accession # NP_062562
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2155
Test Specificity Detects human Coagulation Factor VII in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) Kallikrein-1, -2, -3, -4, -5, -7, -8, -9, -10, -11, -12, -13, -14, -15, -B1, rhHGFA, rhFactor X, rhFactor XI, rhThrombin, rhuPA, or rhPROC is observed.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2b
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.