Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Coagulation Factor VII Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB23381
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB23381 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB23381 Supplier R&D Systems Supplier No. MAB23381
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Coagulation Factor VII Monoclonal specifically detects Coagulation Factor VII in Human samples. It is validated for Flow Cytometry, CyTOF-ready.

Specifications

Antigen Coagulation Factor VII
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 321605
Concentration LYOPH
Conjugate Unconjugated
Dilution Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. NP_062562
Gene Alias coagulation factor VII, coagulation factor VII (serum prothrombin conversion accelerator), EC 3.4.21, EC 3.4.21.21, Eptacog alfa, F7, FVII coagulation protein, Proconvertin, Serum prothrombin conversion accelerator, SPCA
Gene Symbols F7
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Coagulation Factor VII Ala39-Pro444 Accession # NP_062562
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2155
Test Specificity Detects human Coagulation Factor VII in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) Kallikrein-1, -2, -3, -4, -5, -7, -8, -9, -10, -11, -12, -13, -14, -15, -B1, rhHGFA, rhFactor X, rhFactor XI, rhThrombin, rhuPA, or rhPROC is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.