Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Coagulation Factor VII Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB2338SP
This item is not returnable.
View return policy
Description
Coagulation Factor VII Monoclonal specifically detects Coagulation Factor VII in Human samples. It is validated for Western Blot, Immunoprecipitation, Immunocytochemistry.Specifications
Coagulation Factor VII | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL, Immunocytochemistry 8-25 ug/mL | |
NP_062562 | |
F7 | |
Mouse myeloma cell line NS0-derived recombinant human Coagulation Factor VII Ala39-Pro444 Accession # NP_062562 | |
25 μg | |
Primary | |
Detects human Coagulation Factor VII in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human (rh) Kallikrein-1, -2, -3, -4, -5, -7, -8, -9, -10, -11, -12, -13, -14, -15, -B1, rhHGFA, rhFactor X, rhFactor XI, rhThrombin, rhuPA-1, or rhPROC is observed. | |
Human | |
Purified |
Western Blot, Immunoprecipitation, Immunocytochemistry | |
321621 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
coagulation factor VII, coagulation factor VII (serum prothrombin conversion accelerator), EC 3.4.21, EC 3.4.21.21, Eptacog alfa, F7, FVII coagulation protein, Proconvertin, Serum prothrombin conversion accelerator, SPCA | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
2155 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction