Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human COCO Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB30471
Change view
Click to view available options
Quantity:
25 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB30471 500 μg
MAB30471SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB30471 Supplier R&D Systems Supplier No. MAB30471
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

COCO/DAND5 Monoclonal specifically detects COCO/DAND5 in Human samples. It is validated for ELISA Capture (Matched Antibody Pair).

Specifications

Antigen COCO/DAND5
Applications ELISA
Classification Monoclonal
Clone 322209
Conjugate Unconjugated
Dilution ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q8N907
Gene Alias CER2CRL2, Cerberus 2, Cerberus like-2, cerberus-like 2, Cerberus-like protein 2, cerl-2, CERL2, CKTSF1B3, CKTSF1B3MGC126849, COCO, Cysteine knot superfamily 1, BMP antagonist 3, DAN domain family member 5, DAN domain family, member 5, DAND5, Dante, DTE, FLJ38607, GREM3, GREM3cerberus 2, Gremlin-3, SP1
Gene Symbols DAND5
Host Species Mouse
Immunogen E. coli-derived recombinant human COCO Arg23-Ala189 Accession # Q8N907
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 199699
Test Specificity Detects human COCO in ELISAs. In sandwich immunoassays, no cross-reactivity or interference with recombinant human (rh) Activin A, rhBMP-4, rhDAN, or recombinant mouse COCO is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.