Learn More
Novus Biologicals™ Human Coenzyme Q10 (CoQ10) ELISA Kit (Colorimetric)
Shop All R&D Systems Products

Specifications
Specifications
| Assay Range | 3.12 - 50 ng/mL |
| Assay Sensitivity | 1.56 ng/mL |
| Product Type | ELISA Kit (Colorimetric) |
| Sample Type | Plasma, Serum, Tissue Homogenates |
| Specificity | This assay has high sensitivity and excellent specificity for detection of human Coenzyme Q10 (CoQ10). No significant cross-reactivity or interference between human Coenzyme Q10 (CoQ10) and analogues was observed. |
| For Use With (Application) | ELISA |
| Quantity | 1 Kit |
| Storage Requirements | Store at 4°C. |
| Target | Human |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.