Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Collagen I alpha 1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB62202100
This item is not returnable.
View return policy
Description
Collagen I alpha 1 Monoclonal antibody specifically detects Collagen I alpha 1 in Human samples. It is validated for ELISASpecifications
Collagen I alpha 1 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
Alpha-1 type I collagen, COL1A1, COL-IA1, Collagen 1, Collagen 1 alpha 1, collagen alpha 1 chain type I, collagen alpha-1(I) chain, collagen of skin, tendon and bone, alpha-1 chain, Collagen Type I Alpha 1 Chain, collagen, type I, alpha 1, Collagen1, Collagen-1, EDSARTH1, OI4, pro-alpha-1 collagen type 1, Type I Procollagen Alpha 1 Chain | |
Chinese Hamster Ovary cell line, CHO-derived human Collagen I alpha 1, Gln23-Lys277 + Gly1094-Leu1464,, Accession # P02452 | |
100 μg | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. | |
IgG2b |
ELISA | |
816154 | |
ELISA | |
P02452 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
1277 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction