Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Collagen IV alpha 1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB6308
This item is not returnable.
View return policy
Description
Collagen IV alpha 1 Monoclonal specifically detects Collagen IV alpha 1 in Human samples. It is validated for Immunocytochemistry.Specifications
Collagen IV alpha 1 | |
Monoclonal | |
LYOPH | |
Immunocytochemistry 8-25 ug/mL | |
P02462 | |
COL4A1 | |
Mouse myeloma cell line NS0-derived recombinant human Collagen IV alpha 1 Ser1441-Thr1669 Accession # P02462 | |
100 μg | |
Primary | |
Detects human Collagen IV alpha 1 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human Collagen I alpha 1, III alpha 1, XIII alpha 1v4, XXIII alpha 1, or recombinant mouse Collagen XXV alpha 1 is observed. | |
Human | |
Purified |
Immunocytochemistry | |
577238 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
alpha-1 chain, arresten, COL4A1, COL4A1 NC1 domain, collagen alpha-1(IV) chain, collagen IV, alpha-1 polypeptide, collagen, type IV, alpha 1, Collagen-4, EC 3.4.23, EC 6.3.1.2 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
1282 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction