Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Human Collagen IV ELISA Kit (Colorimetric)

Click to view available options
Quantity:
1 Kit
Description
The Human Collagen IV ELISA Kit (Colorimetric) measures human Collagen IV in serum, plasma and other biological fluids.
Specifications
Specifications
Assay Range | 0.78 - 50 ng/mL |
Assay Sensitivity | 0.47 ng/mL |
Cross Reactivity | This kit recognizes Human COL4 in samples. No significant cross-reactivity or interference between Human COL4 and analogues was observed. |
Product Type | ELISA Kit (Colorimetric) |
Sample Type | Biological Fluids, Plasma, Serum |
For Use With (Application) | ELISA |
Host Species | Human |
Quantity | 1 Kit |
Storage Requirements | Storage of components varies. See protocol for specific instructions. |
Target | Collagen IV |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction